Please login to your online account to display your discounted pricing

1-Bromo-3,5-difluorobenzene, 99%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  249390100

 View more versions of this product

Catalog No. AC249390100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 192.99
Quantity 10mL
Color Clear colorless to light yellow
Beilstein 05, 679
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula BrC6H3F2
Refractive Index 1.4980 to 1.5000 (20°C, 589nm)
Chemical Name or Material 1-Bromo-3, 5-difluorobenzene
Molecular Formula C6H3BrF2
Assay Percent Range 99%
CAS 461-96-1
Health Hazard 1 Warning
MDL Number MFCD00010305
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

Specifications

Chemical Name or Material 1-Bromo-3, 5-difluorobenzene
CAS 461-96-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3BrF2
Linear Formula BrC6H3F2
Formula Weight 192.99
MDL Number MFCD00010305
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

SDS & Safety Documents