missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3,5-difluorobenzene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 249390025

 View more versions of this product

Catalog No. AC249390025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Bromo-3, 5-difluorobenzene
45°C
Glass bottle
1.676
Colorless to Yellow
2.5mL
98.5
99%
BrC6H3F2
05, 679
Solubility in water: 0.238g/L (20°C)
FC1=CC(Br)=CC(F)=C1
192.99
192.99
99%
1.6760g/mL
Authentic
1.4980 to 1.5000
140.0°C
-27.0°C
461-96-1
100.0
C6H3BrF2
MFCD00010305
1-bromo-3,5-difluoro benzene, 1-bromo-3,5-difluoro-benzene, 3,5-difluoro bromobenzene, 3,5-difluoro-bromobenzene, 3,5-difluorobromobenzene, 3,5-difluorophenyl bromide, acmc-1ae0k, benzene, 1-bromo-3,5-difluoro, bromo-3,5-difluorobenzene, pubchem1042
JHLKSIOJYMGSMB-UHFFFAOYSA-N
1-bromo-3,5-difluorobenzene
136313
Liquid

Chemical Identifiers

461-96-1
192.99
JHLKSIOJYMGSMB-UHFFFAOYSA-N
136313
FC1=CC(Br)=CC(F)=C1
C6H3BrF2
MFCD00010305
1-bromo-3,5-difluoro benzene, 1-bromo-3,5-difluoro-benzene, 3,5-difluoro bromobenzene, 3,5-difluoro-bromobenzene, 3,5-difluorobromobenzene, 3,5-difluorophenyl bromide, acmc-1ae0k, benzene, 1-bromo-3,5-difluoro, bromo-3,5-difluorobenzene, pubchem1042
1-bromo-3,5-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents
Product Certifications