missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3,5-difluorobenzene, 98%

$138.68 - $138.68

Chemical Identifiers

CAS 461-96-1
Molecular Formula C6H3BrF2
Formula Weight 192.99
MDL Number MFCD00010305
UN Number UN1993
Beilstein 1931520
Synonym 3,5-Difluorobromobenzene
Chemical Name or Material 1-Bromo-3,5-difluorobenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1405314 Alfa Aesar™
A1405314
25g Each for $138.68
AAA1405306 Alfa Aesar™
A1405306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1405322 Alfa Aesar™
A1405322
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1405330 Alfa Aesar™
A1405330
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

461-96-1
192.99
UN1993
3,5-Difluorobromobenzene
C6H3BrF2
MFCD00010305
1931520
1-Bromo-3,5-difluorobenzene

Specifications

461-96-1
192.99
UN1993
1931520
140° to 141°C
44°C (111°F)
98%
C6H3BrF2
MFCD00010305
−27°C
3,5-Difluorobromobenzene
25g
1-Bromo-3,5-difluorobenzene

Safety and Handling

GHS H Statement
H226-H373-H302-H315-H317
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P261-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.