missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3,5-dichlorobenzene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 331901000

 View more versions of this product

Catalog No. AC331901000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Powder
White to beige-brown
100.0
98%
19752-55-7
MFCD00000584
Authentic
225.9
100g
74° to 77°C
C6H3BrCl2
98%
1-Bromo-3, 5-dichlorobenzene
97.5
Glass bottle

Chemical Identifiers

19752-55-7
100.0
C6H3BrCl2
MFCD00000584
≥97.5%
97.5
98%
225.9
1-Bromo-3, 5-dichlorobenzene, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-270-7

TSCA : TSCA