missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3,4-difluorobenzene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 233190100

 View more versions of this product

Catalog No. AC233190100



Specifications

Specifications

1.7070g/mL
Authentic
1.5040 to 1.506
150°C to 151°C
-4°C
98%
97.5
97.5% min. (GC)
BrC6H3F2
1-bromo-3,4-difluorobenzene, 3,4-difluoro-1-bromobenzene, 3,4-difluorobromebenzene, 3,4-difluorobromobenzene, 3,4-difluorophenylbromide, 4-bromo-1,2-difluoro-benzene, benzene, 4-bromo-1,2-difluoro, fr bf de, paragos 390209, pubchem2303
YMQPKONILWWJQG-UHFFFAOYSA-N
4-bromo-1,2-difluorobenzene
67675
Liquid
33°C
Glass bottle
1.707
Colorless to Yellow
10g
348-61-8
100.0
C6H3BrF2
MFCD00000304
Solubility in water: insoluble
C1=CC(=C(C=C1Br)F)F
193
193
98%

Chemical Identifiers

348-61-8
193
YMQPKONILWWJQG-UHFFFAOYSA-N
67675
C1=CC(=C(C=C1Br)F)F
C6H3BrF2
MFCD00000304
1-bromo-3,4-difluorobenzene, 3,4-difluoro-1-bromobenzene, 3,4-difluorobromebenzene, 3,4-difluorobromobenzene, 3,4-difluorophenylbromide, 4-bromo-1,2-difluoro-benzene, benzene, 4-bromo-1,2-difluoro, fr bf de, paragos 390209, pubchem2303
4-bromo-1,2-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-481-5

TSCA : TSCA

SDS Documents