Learn More
1-Bromo-3,4-difluorobenzene, 98%, ACROS Organics™
$106.95 - $106.95
Chemical Identifiers
CAS | 348-61-8 |
---|---|
Molecular Formula | C6H3BrF2 |
Molecular Weight (g/mol) | 193 |
MDL Number | MFCD00000304 |
InChI Key | YMQPKONILWWJQG-UHFFFAOYSA-N |
Synonym | 1-bromo-3,4-difluorobenzene, 3,4-difluoro-1-bromobenzene, 3,4-difluorobromebenzene, 3,4-difluorobromobenzene, 3,4-difluorophenylbromide, 4-bromo-1,2-difluoro-benzene, benzene, 4-bromo-1,2-difluoro, fr bf de, paragos 390209, pubchem2303 |
PubChem CID | 67675 |
IUPAC Name | 4-bromo-1,2-difluorobenzene |
SMILES | C1=CC(=C(C=C1Br)F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC233190010
|
Thermo Fisher Scientific
233190010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC233190100
|
Thermo Fisher Scientific
233190100 |
10g | Glass bottle |
Each for $106.95
|
|
Chemical Identifiers
348-61-8 | |
193 | |
YMQPKONILWWJQG-UHFFFAOYSA-N | |
67675 | |
C1=CC(=C(C=C1Br)F)F |
C6H3BrF2 | |
MFCD00000304 | |
1-bromo-3,4-difluorobenzene, 3,4-difluoro-1-bromobenzene, 3,4-difluorobromebenzene, 3,4-difluorobromobenzene, 3,4-difluorophenylbromide, 4-bromo-1,2-difluoro-benzene, benzene, 4-bromo-1,2-difluoro, fr bf de, paragos 390209, pubchem2303 | |
4-bromo-1,2-difluorobenzene |
Specifications
1.7070g/mL | |
Authentic | |
1.5040 to 1.506 | |
150°C to 151°C | |
-4°C | |
348-61-8 | |
100.0 | |
C6H3BrF2 | |
MFCD00000304 | |
Solubility in water: insoluble | |
C1=CC(=C(C=C1Br)F)F | |
193 | |
193 | |
98% |
33°C | |
Glass bottle | |
1.707 | |
Colorless to Yellow | |
1g | |
97.5 | |
97.5% min. (GC) | |
BrC6H3F2 | |
1-bromo-3,4-difluorobenzene, 3,4-difluoro-1-bromobenzene, 3,4-difluorobromebenzene, 3,4-difluorobromobenzene, 3,4-difluorophenylbromide, 4-bromo-1,2-difluoro-benzene, benzene, 4-bromo-1,2-difluoro, fr bf de, paragos 390209, pubchem2303 | |
YMQPKONILWWJQG-UHFFFAOYSA-N | |
4-bromo-1,2-difluorobenzene | |
67675 | |
Liquid | |
1-Bromo-3, 4-difluorobenzene, 98% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 206-481-5
TSCA : TSCA