missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3,3-diphenylpropane, 99%, ACROS Organics™

Chemical Identifiers

CAS 20017-68-9
Molecular Formula C15H15Br
Molecular Weight (g/mol) 275.189
MDL Number MFCD00044829
InChI Key SLHSRCBFPHCSGL-UHFFFAOYSA-N
Synonym 1-bromo-3,3-diphenylpropane, 3,3-diphenylpropyl bromide, 3-bromo-1,1-diphenylpropane, 3-bromopropane-1,1-diyl dibenzene, 3-bromo-1-phenylpropyl benzene, 3,3-diphenylpropylbromide, 1,1'-3-bromopropylidene bisbenzene, benzene, 1,1'-3-bromopropylidene bis, acmc-1cjoi, 3,3-diphenyl-1-propylbromide
PubChem CID 88333
IUPAC Name (3-bromo-1-phenylpropyl)benzene
SMILES C1=CC=C(C=C1)C(CCBr)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC136110010 SDS View Documents Acros Organics
136110010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

20017-68-9
275.189
SLHSRCBFPHCSGL-UHFFFAOYSA-N
88333
C1=CC=C(C=C1)C(CCBr)C2=CC=CC=C2
C15H15Br
MFCD00044829
1-bromo-3,3-diphenylpropane, 3,3-diphenylpropyl bromide, 3-bromo-1,1-diphenylpropane, 3-bromopropane-1,1-diyl dibenzene, 3-bromo-1-phenylpropyl benzene, 3,3-diphenylpropylbromide, 1,1'-3-bromopropylidene bisbenzene, benzene, 1,1'-3-bromopropylidene bis, acmc-1cjoi, 3,3-diphenyl-1-propylbromide
(3-bromo-1-phenylpropyl)benzene

Specifications

Authentic
White
1g
98.5
99%
(C6H5)2CH(CH2)2Br
1-bromo-3,3-diphenylpropane, 3,3-diphenylpropyl bromide, 3-bromo-1,1-diphenylpropane, 3-bromopropane-1,1-diyl dibenzene, 3-bromo-1-phenylpropyl benzene, 3,3-diphenylpropylbromide, 1,1'-3-bromopropylidene bisbenzene, benzene, 1,1'-3-bromopropylidene bis, acmc-1cjoi, 3,3-diphenyl-1-propylbromide
C1=CC=C(C=C1)C(CCBr)C2=CC=CC=C2
275.189
275.19
99%
Glass bottle
39.0°C to 42.0°C
20017-68-9
100.0
C15H15Br
MFCD00044829
SLHSRCBFPHCSGL-UHFFFAOYSA-N
(3-bromo-1-phenylpropyl)benzene
88333
Crystalline Powder
1-Bromo-3, 3-diphenylpropane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger