Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3,3-diphenylpropane, 99%

$105.89 - $474.09

Chemical Identifiers

Chemical Name or Material 1-Bromo-3,3-diphenylpropane
CAS 20017-68-9
Molecular Formula C15H15Br
Formula Weight 275.18
MDL Number MFCD00044829
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2555203

  SDS
alfa aesar™
B2555203
1g Each for $105.89

AAB2555206

  SDS
alfa aesar™
B2555206
5g Each for $474.09
Description & Specifications

Specifications

Formula Weight 275.18
Chemical Name or Material 1-Bromo-3,3-diphenylpropane
Assay 99%
Molecular Formula C15H15Br
MDL Number MFCD00044829
CAS 20017-68-9
Melting Point 39° to 42°C
Quantity 1g
Odor Odorless