Please login to your online account to display your discounted pricing

1-Bromo-3,3-dimethylbutan-2-one, Tech., Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CD08380EA

 View more versions of this product

Catalog No. CD08380EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 179.06
Quantity 10g
Assay Tech.
Density 1.3300g/mL
CAS Max % 100.0
CAS Min % 80.0
Synonym 1-Bromo-3, 3-dimethyl-2-butanone
Chemical Name or Material 1-Bromo-3, 3-dimethylbutan-2-one
CAS 5469-26-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00000206
Packaging Amber glass bottle
Linear Formula (CH3)3CCOCH2Br
Molecular Formula C6H11BrO
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 1-Bromo-3, 3-dimethylbutan-2-one
CAS 5469-26-1
CAS Min % 80.0
CAS Max % 100.0
Assay Tech.
Molecular Formula C6H11BrO
Linear Formula (CH3)3CCOCH2Br
Formula Weight 179.06
MDL Number MFCD00000206
Synonym 1-Bromo-3, 3-dimethyl-2-butanone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents