missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3,3,4,4,4-pentafluoro-2-butanone, 97%

Manufacturer:  Alfa Aesar™ L1219906

 View more versions of this product

Catalog No. AAL1219906



Specifications

Specifications

1-Bromo-3,3,4,4,4-pentafluoro-2-butanone
5g
C4H2BrF5O
UN3265
HHGIRNBWKHUFNA-UHFFFAOYSA-N
1-bromo-3,3,4,4,4-pentafluorobutan-2-one
2736355
97%
93°C to 94°C
92737-01-4
MFCD00041447
1-bromo-3,3,4,4,4-pentafluoro-2-butanone, 2-butanone, 1-bromo-3,3,4,4,4-pentafluoro, 1-bromo-3,3,4,4,4-pentafluorobutanone, 2-butanone,1-bromo-3,3,4,4,4-pentafluoro, 1-bromanyl-3,3,4,4,4-pentakis fluoranyl butan-2-one
C(C(=O)C(C(F)(F)F)(F)F)Br
240.955
240.96

Chemical Identifiers

92737-01-4
240.955
HHGIRNBWKHUFNA-UHFFFAOYSA-N
2736355
C(C(=O)C(C(F)(F)F)(F)F)Br
C4H2BrF5O
MFCD00041447
1-bromo-3,3,4,4,4-pentafluoro-2-butanone, 2-butanone, 1-bromo-3,3,4,4,4-pentafluoro, 1-bromo-3,3,4,4,4-pentafluorobutanone, 2-butanone,1-bromo-3,3,4,4,4-pentafluoro, 1-bromanyl-3,3,4,4,4-pentakis fluoranyl butan-2-one
1-bromo-3,3,4,4,4-pentafluorobutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents