missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-3,3,3-trifluoroacetone, 97%

Manufacturer:  Alfa Aesar™ A1494822

 View more versions of this product

Catalog No. AAA1494822



Description

Description

Applications
1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

1-Bromo-3,3,3-trifluoroacetone
Pungent
<-80°C
431-35-6
MFCD00039237
1703387
ONZQYZKCUHFORE-UHFFFAOYSA-N
3-bromo-1,1,1-trifluoropropan-2-one
79008
97%
−2°C (28°F)
85°C to 87°C
100g
C3H2BrF3O
UN2924
3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
C(C(=O)C(F)(F)F)Br
190.947
190.95

Chemical Identifiers

431-35-6
190.947
ONZQYZKCUHFORE-UHFFFAOYSA-N
79008
C(C(=O)C(F)(F)F)Br
C3H2BrF3O
MFCD00039237
3-bromo-1,1,1-trifluoroacetone, 1-bromo-3,3,3-trifluoroacetone, bromotrifluoroacetone, 3-bromo-1,1,1-trifluoro-2-propanone, 3-bromo-1,1,1-trifluoropropanone, 1-bromo-3,3,3-trifluoro-2-propanone, 2-propanone, 3-bromo-1,1,1-trifluoro, 1,1,1-trifluoro-3-bromopropanone, 3-bromo-1,1,1-trifluoro-propan-2-one, bromotrifloroacetone
3-bromo-1,1,1-trifluoropropan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Documents