missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(trifluoromethoxy)benzene 98.0+%, TCI America™

Supplier:  TCI America T214725G

 View more versions of this product

Catalog No. T214725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Bromo-2-(trifluoromethoxy)benzene
Yellow
C7H4BrF3O
25 g
1-bromo-2-trifluoromethoxy benzene, 2-trifluoromethoxy bromobenzene, 2-bromo trifluoromethoxy benzene, o-trifluoromethoxybromobenzene, o-bromo trifluoromethoxy benzene, 2-bromo-alpha,alpha,alpha-trifluoroanisole, 2-bromophenyl trifluoromethyl ether, trifluoromethoxy bromobenzene, o-bromotrifluoromethoxybenzene, benzene, bromo trifluoromethoxy
FC(F)(F)OC1=CC=CC=C1Br
241.01
241.01
Liquid
64115-88-4
160°C
MFCD00040942
1993
ITYCJCVRPBLODP-UHFFFAOYSA-N
1-bromo-2-(trifluoromethoxy)benzene
2777266
≥98.0% (GC)

Chemical Identifiers

64115-88-4
241.01
ITYCJCVRPBLODP-UHFFFAOYSA-N
2777266
FC(F)(F)OC1=CC=CC=C1Br
C7H4BrF3O
MFCD00040942
1-bromo-2-trifluoromethoxy benzene, 2-trifluoromethoxy bromobenzene, 2-bromo trifluoromethoxy benzene, o-trifluoromethoxybromobenzene, o-bromo trifluoromethoxy benzene, 2-bromo-alpha,alpha,alpha-trifluoroanisole, 2-bromophenyl trifluoromethyl ether, trifluoromethoxy bromobenzene, o-bromotrifluoromethoxybenzene, benzene, bromo trifluoromethoxy
1-bromo-2-(trifluoromethoxy)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions