missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-(trifluoromethoxy)benzene 98.0+%, TCI America™
$33.56 - $61.51
Chemical Identifiers
| CAS | 64115-88-4 |
|---|---|
| Molecular Formula | C7H4BrF3O |
| Molecular Weight (g/mol) | 241.01 |
| MDL Number | MFCD00040942 |
| InChI Key | ITYCJCVRPBLODP-UHFFFAOYSA-N |
| Synonym | 1-bromo-2-trifluoromethoxy benzene, 2-trifluoromethoxy bromobenzene, 2-bromo trifluoromethoxy benzene, o-trifluoromethoxybromobenzene, o-bromo trifluoromethoxy benzene, 2-bromo-alpha,alpha,alpha-trifluoroanisole, 2-bromophenyl trifluoromethyl ether, trifluoromethoxy bromobenzene, o-bromotrifluoromethoxybenzene, benzene, bromo trifluoromethoxy |
| PubChem CID | 2777266 |
| IUPAC Name | 1-bromo-2-(trifluoromethoxy)benzene |
| SMILES | FC(F)(F)OC1=CC=CC=C1Br |
Chemical Identifiers
| 64115-88-4 | |
| 241.01 | |
| ITYCJCVRPBLODP-UHFFFAOYSA-N | |
| 2777266 | |
| FC(F)(F)OC1=CC=CC=C1Br |
| C7H4BrF3O | |
| MFCD00040942 | |
| 1-bromo-2-trifluoromethoxy benzene, 2-trifluoromethoxy bromobenzene, 2-bromo trifluoromethoxy benzene, o-trifluoromethoxybromobenzene, o-bromo trifluoromethoxy benzene, 2-bromo-alpha,alpha,alpha-trifluoroanisole, 2-bromophenyl trifluoromethyl ether, trifluoromethoxy bromobenzene, o-bromotrifluoromethoxybenzene, benzene, bromo trifluoromethoxy | |
| 1-bromo-2-(trifluoromethoxy)benzene |
Specifications
| 64115-88-4 | |
| 160°C | |
| MFCD00040942 | |
| 1993 | |
| ITYCJCVRPBLODP-UHFFFAOYSA-N | |
| 1-bromo-2-(trifluoromethoxy)benzene | |
| 2777266 | |
| ≥98.0% (GC) | |
| 1-Bromo-2-(trifluoromethoxy)benzene |
| Yellow | |
| C7H4BrF3O | |
| 5 g | |
| 1-bromo-2-trifluoromethoxy benzene, 2-trifluoromethoxy bromobenzene, 2-bromo trifluoromethoxy benzene, o-trifluoromethoxybromobenzene, o-bromo trifluoromethoxy benzene, 2-bromo-alpha,alpha,alpha-trifluoroanisole, 2-bromophenyl trifluoromethyl ether, trifluoromethoxy bromobenzene, o-bromotrifluoromethoxybenzene, benzene, bromo trifluoromethoxy | |
| FC(F)(F)OC1=CC=CC=C1Br | |
| 241.01 | |
| 241.01 | |
| Liquid |
Safety and Handling
TSCA : No