Please login to your online account to display your discounted pricing

1-Bromo-2-propanol, 70%, remainder 2-bromo-1-propanol, tech., ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC218620500

 View more versions of this product

Catalog No. AC218620500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥65%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 138.99
Quantity 50mL
Color Clear colorless to brown
Density 1.5300g/mL
Beilstein 01, 365
CAS Min % 65.0
CAS Max % 100.0
Assay Percent Range 70%
Molecular Formula C3H7BrO
Linear Formula CH3CH(OH)CH2Br
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00004529
CAS 598-18-5
Chemical Name or Material 1-Bromo-2-propanol
CAS 19686-73-8
Health Hazard 1 Warning
Infrared Spectrum Authentic
Refractive Index 1.4790 to 1.4900 (20°C, 589nm)

Specifications

Chemical Name or Material 1-Bromo-2-propanol
CAS 19686-73-8
CAS Min % 65.0
CAS Max % 100.0
CAS 598-18-5
Purity Grade Technical Grade
Assay ≥65%
Assay Percent Range 70%
Molecular Formula C3H7BrO
Linear Formula CH3CH(OH)CH2Br
Formula Weight 138.99
MDL Number MFCD00004529
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents