missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-propanol, 70%, remainder 2-bromo-1-propanol, tech., ACROS Organics™

$44.03 - $44.03

Chemical Identifiers

CAS 19686-73-8,598-18-5
CAS Min % 65.0
CAS Max % 100.0
Assay Percent Range 70%
Molecular Formula C3H7BrO
Linear Formula CH3CH(OH)CH2Br
Formula Weight 138.99
MDL Number MFCD00004529
Chemical Name or Material 1-Bromo-2-propanol
Assay ≥65%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC218620500 SDS Thermo Fisher Scientific
AC218620500
Glass bottle 50mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC218620100 SDS Thermo Fisher Scientific
218620100
Glass bottle 10mL Each for $44.03
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

19686-73-8,598-18-5
100.0
C3H7BrO
138.99
1-Bromo-2-propanol
Technical Grade
65.0
70%
CH3CH(OH)CH2Br
MFCD00004529
≥65%

Specifications

19686-73-8,598-18-5
Authentic
Glass bottle
1.5300g/mL
Liquid
70%
CH3CH(OH)CH2Br
MFCD00004529
1-Bromo-2-propanol
Technical Grade
65.0
100.0
1.4790 to 1.4900 (20°C, 589nm)
50mL
Clear colorless to brown
C3H7BrO
138.99
01, 365
≥65%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning