Please login to your online account to display your discounted pricing

1-Bromo-2-propanol, 70%, remainder 2-bromo-1-propanol, tech., ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  218620100

 View more versions of this product

Catalog No. AC218620100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥65%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 138.99
Quantity 10mL
Color Clear colorless to brown
Beilstein 01, 365
CAS Max % 100.0
CAS Min % 65.0
Assay Percent Range 70%
Linear Formula CH3CH(OH)CH2Br
CAS 19686-73-8
Infrared Spectrum Authentic
Refractive Index 1.4790 to 1.4900 (20°C, 589nm)
Chemical Name or Material 1-Bromo-2-propanol
Molecular Formula C3H7BrO
MDL Number MFCD00004529
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
CAS 598-18-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 1-Bromo-2-propanol
CAS 19686-73-8
CAS Min % 65.0
CAS Max % 100.0
CAS 598-18-5
Purity Grade Technical Grade
Assay ≥65%
Assay Percent Range 70%
Molecular Formula C3H7BrO
Linear Formula CH3CH(OH)CH2Br
Formula Weight 138.99
MDL Number MFCD00004529
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.