Learn More
1-Bromo-2-nitrobenzene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1168630
Description
1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2?-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-2-nitrobenzene is used in organic synthesis. It also can react with 2-methylamino-benzoic acid to get N-methyl-N-(2-nitro-phenyl)-anthranilic acid. And it is also used in the preparation of 4-methoxy-2′-nitrodiphenyl ether, 1-methoxy-3,5-bis-(2-nitro-phenoxy)benzene.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
1-Bromo-2-nitrobenzene | |
40°C to 44°C | |
112°C (233°F) | |
MFCD00007045 | |
UN3459 | |
2-bromonitrobenzene, o-bromonitrobenzene, o-nitrobromobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, o-nitrophenyl bromide, 2-bromo-1-nitrobenzene, 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, benzene, bromonitro | |
ORPVVAKYSXQCJI-UHFFFAOYSA-N | |
1-bromo-2-nitrobenzene | |
11341 | |
99% |
577-19-5 | |
261°C | |
C6H4BrNO2 | |
250 g | |
2044109 | |
Insoluble in water. | |
C1=CC=C(C(=C1)[N+](=O)[O-])Br | |
202.007 | |
202.01 |
Chemical Identifiers
577-19-5 | |
202.007 | |
ORPVVAKYSXQCJI-UHFFFAOYSA-N | |
11341 | |
C1=CC=C(C(=C1)[N+](=O)[O-])Br |
C6H4BrNO2 | |
MFCD00007045 | |
2-bromonitrobenzene, o-bromonitrobenzene, o-nitrobromobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, o-nitrophenyl bromide, 2-bromo-1-nitrobenzene, 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, benzene, bromonitro | |
1-bromo-2-nitrobenzene |
Safety and Handling
GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROBROMOBENZENES, SOLID
EINECSNumber : 209-409-0
RTECSNumber : CY9040450
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only