missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-nitrobenzene, 98%, ACROS Organics™

Chemical Identifiers

CAS 577-19-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00007045
InChI Key ORPVVAKYSXQCJI-UHFFFAOYSA-N
Synonym 1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, 2-bromo-1-nitrobenzene, 2-bromonitrobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, benzene, bromonitro, o-bromonitrobenzene, o-nitrobromobenzene, o-nitrophenyl bromide
PubChem CID 11341
IUPAC Name 1-bromo-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC150350050
SDS View Documents
Thermo Fisher Scientific
AC150350050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

577-19-5
202.01
ORPVVAKYSXQCJI-UHFFFAOYSA-N
11341
C1=CC=C(C(=C1)[N+](=O)[O-])Br
C6H4BrNO2
MFCD00007045
1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, 2-bromo-1-nitrobenzene, 2-bromonitrobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, benzene, bromonitro, o-bromonitrobenzene, o-nitrobromobenzene, o-nitrophenyl bromide
1-bromo-2-nitrobenzene

Specifications

>110°C
261.0°C
5g
97.5
97.5% min. (GC)
BrC6H4NO2
05, I, 247
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
C1=CC=C(C(=C1)[N+](=O)[O-])Br
202.01
202.01
1-Bromo-2-nitrobenzene, 98%
Glass bottle
40.0°C to 43.0°C
577-19-5
100.0
C6H4BrNO2
MFCD00007045
1-bromo-2-nitro-benzene, 2-bromo nitrobenzene, 2-bromo-1-nitrobenzene, 2-bromonitrobenzene, 2-nitrobromobenzene, benzene, 1-bromo-2-nitro, benzene, bromonitro, o-bromonitrobenzene, o-nitrobromobenzene, o-nitrophenyl bromide
ORPVVAKYSXQCJI-UHFFFAOYSA-N
1-bromo-2-nitrobenzene
11341
98%
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause damage to organs through

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia

GHS Signal Word: Warning

EINECSNumber : 209-409-0

RTECSNumber : CY9040450

TSCA : TSCA

SDS