Please login to your online account to display your discounted pricing

1-Bromo-2-naphthol 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107161000

 View more versions of this product

Catalog No. AC107161000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 223.07
Quantity 100g
Color Off-white to beige or brown-purple
EINECS Number 209-363-1
TSCA TSCA
Melting Point 77° to 81°C
RTECS Number QL3361000
Beilstein 06, 650
CAS Max % 100.0
Chemical Name or Material 1-Bromo-2-naphthol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H7BrO
Linear Formula BrC10H6OH
MDL Number MFCD00003869
CAS 573-97-7
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 1-Bromo-2-naphthol
Name Note 98%
CAS 573-97-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H7BrO
Linear Formula BrC10H6OH
Formula Weight 223.07
MDL Number MFCD00003869
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents