missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-naphthol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107160250

 View more versions of this product

Catalog No. AC107160250



Description

Description

Specifications

Specifications

1-Bromo-2-naphthol
Glass bottle
77.0°C to 81.0°C
98%
97.5
97.5% min. (GC)
BrC10H6OH
06, 650
Solubility in water: insoluble
C1=CC=C2C(=C1)C=CC(=C2Br)O
223.07
223.07
98%
Authentic
Beige to White or Brown-Purple
25g
573-97-7
100.0
C10H7BrO
MFCD00003869
1-brom-2-naphthol, 1-brom-2-naphthol german, 1-bromo-2-hydroxynaphthalene, 1-bromo-2-naphthalenol, 1-bromo-2-naphthol, 2-naphthalenol, 1-bromo, 2-naphthol, 1-bromo, disthemin, unii-vke7l55pp9, wormin
FQJZPYXGPYJJIH-UHFFFAOYSA-N
1-bromonaphthalen-2-ol
11316
Crystalline Powder

Chemical Identifiers

573-97-7
223.07
FQJZPYXGPYJJIH-UHFFFAOYSA-N
11316
C1=CC=C2C(=C1)C=CC(=C2Br)O
C10H7BrO
MFCD00003869
1-brom-2-naphthol, 1-brom-2-naphthol german, 1-bromo-2-hydroxynaphthalene, 1-bromo-2-naphthalenol, 1-bromo-2-naphthol, 2-naphthalenol, 1-bromo, 2-naphthol, 1-bromo, disthemin, unii-vke7l55pp9, wormin
1-bromonaphthalen-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 209-363-1

RTECSNumber : QL3361000

TSCA : TSCA

SDS Documents