missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-naphthoic Acid, TCI America™

$48.89 - $785.30

Chemical Identifiers

CAS 20717-79-7
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD00021408
InChI Key VUVIRKAVBZITDO-UHFFFAOYSA-N
Synonym 1-bromo-2-naphthoic acid, 1-bromo-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylic acid, 1-bromo, maybridge4_002377, 1-bromo-2-naphthoicacid, 1-bromo-2-napthoic acid, 1-bromo-2-naphtoic acid, acmc-1ck28, 2-?naphthalenecarboxyli?c acid, 2-naphthalenecarboxylicacid,1-bromo
PubChem CID 88665
IUPAC Name 1-bromonaphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2Br)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16175G View Documents TCI America
B16175G
5g Each for $785.30
 
B16171G View Documents TCI America
B16171G
1g Each for $239.86
 
B1617100MG View Documents TCI America
B1617100MG
100mg Each for $48.89
 
Specifications

Chemical Identifiers

20717-79-7
251.079
VUVIRKAVBZITDO-UHFFFAOYSA-N
88665
C1=CC=C2C(=C1)C=CC(=C2Br)C(=O)O
C11H7BrO2
MFCD00021408
1-bromo-2-naphthoic acid, 1-bromo-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylic acid, 1-bromo, maybridge4_002377, 1-bromo-2-naphthoicacid, 1-bromo-2-napthoic acid, 1-bromo-2-naphtoic acid, acmc-1ck28, 2-?naphthalenecarboxyli?c acid, 2-naphthalenecarboxylicacid,1-bromo
1-bromonaphthalene-2-carboxylic acid

Specifications

White-Yellow
5g
C11H7BrO2
1-bromo-2-naphthoic acid, 1-bromo-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylic acid, 1-bromo, maybridge4_002377, 1-bromo-2-naphthoicacid, 1-bromo-2-napthoic acid, 1-bromo-2-naphtoic acid, acmc-1ck28, 2-?naphthalenecarboxyli?c acid, 2-naphthalenecarboxylicacid,1-bromo
C1=CC=C2C(=C1)C=CC(=C2Br)C(=O)O
251.079
251.08
≥98.0% (GC,T)
191°C
20717-79-7
MFCD00021408
VUVIRKAVBZITDO-UHFFFAOYSA-N
1-bromonaphthalene-2-carboxylic acid
88665
Crystal-Powder at 20°C
1-Bromo-2-naphthoic Acid
Safety and Handling

Safety and Handling

TSCA : No