Please login to your online account to display your discounted pricing

1-Bromo-2-methylpropane, 98+%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC168831000

 View more versions of this product

Catalog No. AC168831000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-methylpropane
CAS 78-77-3
Health Hazard 1 Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4340 to 1.4370 (20°C, 589nm)
Water 0.1% max.
Density 1.2600g/mL
Color Clear colorless
Physical Form Liquid
Quantity 100mL
Assay ≥98.0% (GC)
Assay Percent Range 98+%
Molecular Formula C4H9Br
Linear Formula (CH3)2CHCH2Br
Formula Weight 137.02
Synonym Isobutyl bromide

Specifications

Chemical Name or Material 1-Bromo-2-methylpropane
Assay ≥98.0% (GC)
Assay Percent Range 98+%
Molecular Formula C4H9Br
Linear Formula (CH3)2CHCH2Br
Formula Weight 137.02
Synonym Isobutyl bromide
Accessories

Danger

SDS & Safety Documents