missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-methylpropane, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 168835000

 View more versions of this product

Catalog No. AC168835000


 

Specifications

Specifications

1-Bromo-2-methylpropane
Authentic
1.26
0.1% max.
90.0°C to 92.0°C
Colorless
18°C
98+%
(CH3)2CHCH2Br
HLVFKOKELQSXIQ-UHFFFAOYSA-N
1-bromo-2-methylpropane
6555
Liquid
78-77-3
Glass bottle
1.4346 to 1.4366
1.2600g/mL
-119.0°C
500mL
≥98%
C4H9Br
Isobutyl bromide
CC(C)CBr
137.02
137.02

Chemical Identifiers

78-77-3
137.02
Isobutyl bromide
C4H9Br
HLVFKOKELQSXIQ-UHFFFAOYSA-N
6555
Safety and Handling

Safety and Handling

Danger

SDS Documents