missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-methylnaphthalene, 90%, technical, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 154260050

 View more versions of this product

Catalog No. AC154260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Bromo-2-methylnaphthalene
2586-62-1
100.0
Yellow
>110°C
C11H9Br
CH3C10H6Br
Glass bottle
1.6480 to 1.6500
.beta.-methyl-.alpha.-bromonaphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methylnaphtalene, 1-bromonaphthalen-2-yl methyl, 2-methyl-1-bromonaphthalene, acmc-1cq62, naphthalene, 1-bromo-2-methyl, zlchem 421
CMIMBQIBIZZZHQ-UHFFFAOYSA-N
1-bromo-2-methylnaphthalene
75754
Liquid
Technical
90-95%
90.0
1.4100g/mL
296.0°C
88% min. (GC)
Authentic
MFCD00003871
05, 568
5g
1.41
CC1=CC=C2C=CC=CC2=C1Br
221.10
221.1
90%

Chemical Identifiers

2586-62-1
221.10
CMIMBQIBIZZZHQ-UHFFFAOYSA-N
75754
CC1=CC=C2C=CC=CC2=C1Br
C11H9Br
MFCD00003871
.beta.-methyl-.alpha.-bromonaphthalene, 1-bromanyl-2-methyl-naphthalene, 1-bromo-2-methyl naphthalene, 1-bromo-2-methyl-naphthalene, 1-bromo-2-methylnaphtalene, 1-bromonaphthalen-2-yl methyl, 2-methyl-1-bromonaphthalene, acmc-1cq62, naphthalene, 1-bromo-2-methyl, zlchem 421
1-bromo-2-methylnaphthalene
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 219-966-1

TSCA : TSCA

SDS Documents

Documents

Product Certifications
Promotions

Promotions

RUO – Research Use Only