missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-methoxymethylnaphthalene, TCI America™

$210.84 - $649.31

Chemical Identifiers

CAS 64689-70-9
Molecular Formula C12H11BrO
Molecular Weight (g/mol) 251.123
MDL Number MFCD00059518
InChI Key ORYVAZPUJFACFR-UHFFFAOYSA-N
PubChem CID 12385765
IUPAC Name 1-bromo-2-(methoxymethyl)naphthalene
SMILES COCC1=C(C2=CC=CC=C2C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B11705G View Documents TCI America
B11705G
5g Each for $210.84
 
B117025G View Documents TCI America
B117025G
25g Each for $649.31
 
Specifications

Chemical Identifiers

64689-70-9
251.123
ORYVAZPUJFACFR-UHFFFAOYSA-N
1-bromo-2-(methoxymethyl)naphthalene
C12H11BrO
MFCD00059518
12385765
COCC1=C(C2=CC=CC=C2C=C1)Br

Specifications

151°C
5g
C12H11BrO
ORYVAZPUJFACFR-UHFFFAOYSA-N
1-bromo-2-(methoxymethyl)naphthalene
12385765
Clear Liquid at 20°C
1-Bromo-2-methoxymethylnaphthalene
Undesignated
64689-70-9
MFCD00059518
COCC1=C(C2=CC=CC=C2C=C1)Br
251.123
251.12
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No