Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-isopropylbenzene, 97%

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-isopropylbenzene
CAS 7073-94-1
Molecular Formula C9H11Br
Formula Weight 199.1
MDL Number MFCD00051567
UN Number UN3082
Beilstein 1857014
Synonym 2-Bromocumene; 2-Isopropylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0094106

  SDS
alfa aesar™
L0094106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0094114

  SDS
alfa aesar™
L0094114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.1
Chemical Name or Material 1-Bromo-2-isopropylbenzene
Molecular Formula C9H11Br
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Synonym 2-Bromocumene; 2-Isopropylbromobenzene
Boiling Point 210°C
MDL Number MFCD00051567
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
UN Number UN3082
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
CAS 7073-94-1
Beilstein 1857014

Applications
1-Bromo-2-isopropylbenzene is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dark and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.