Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper

$103.35 - $1,045.69

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-iodotetrafluoroethane
CAS 421-70-5
Name Note stabilized with copper
Molecular Formula BrCF2CF2I
Formula Weight 306.82
MDL Number MFCD00042136
Beilstein 1688466
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2177106

  SDS
alfa aesar™
B2177106
5g Each for $103.35

AAB2177122

  SDS
alfa aesar™
B2177122
100g Each for $1,045.69
Description & Specifications

Specifications

Formula Weight 306.82
Molecular Formula BrCF2CF2I
Recommended Storage 2° to 8°C
Boiling Point 78°C
Name Note stabilized with copper
Beilstein 1688466
Chemical Name or Material 1-Bromo-2-iodotetrafluoroethane
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 421-70-5
Assay 97%
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD00042136
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C