Please login to your online account to display your discounted pricing

1-Bromo-2-iodobenzene, 99%, stabilized, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC221950010

 View more versions of this product

Catalog No. AC221950010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 282.9
Quantity 1g
Color Clear yellow
Beilstein 05, 223
Fieser 01,77
Density 2.2030g/mL
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6H4BrI
MDL Number MFCD00001030
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.6620 to 1.6640 (20°C, 589nm)
Chemical Name or Material 1-Bromo-2-iodobenzene
Linear Formula BrC6H4I
CAS 583-55-1
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Glass bottle
Stabilizer copper

Specifications

Chemical Name or Material 1-Bromo-2-iodobenzene
CAS 583-55-1
CAS Min % 98.5
CAS Max % 100.0
Physical Form Liquid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4BrI
Linear Formula BrC6H4I
Formula Weight 282.9
MDL Number MFCD00001030
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents