missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene, 98%

Manufacturer:  Alfa Aesar™ H6177114

 View more versions of this product

Catalog No. AAH6177114



Specifications

Specifications

1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene
25g
C7H3BrF4O
1-bromo-2-fluoro-4-trifluoromethoxy benzene, 4-bromo-3-fluoro trifluoromethoxy benzene, 3-fluoro-4-bromobenzenetrifluoromethoxy, 4-bromo-3-fluoro-trifluoromethoxy benzene, benzene, 1-bromo-2-fluoro-4-trifluoromethoxy, pubchem1075, acmc-1c50f, ksc498c3b, 4-bromo-3-fluorotrifluoromethoxy-benzene, 2-fluoro-4-trifluoromethoxy bromobenzene
C1=CC(=C(C=C1OC(F)(F)F)F)Br
258.998
259
98%
Yellow
168971-68-4
MFCD06657548
KZMHSGBETSENAT-UHFFFAOYSA-N
1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
22065211
Liquid

Chemical Identifiers

168971-68-4
258.998
KZMHSGBETSENAT-UHFFFAOYSA-N
22065211
C1=CC(=C(C=C1OC(F)(F)F)F)Br
C7H3BrF4O
MFCD06657548
1-bromo-2-fluoro-4-trifluoromethoxy benzene, 4-bromo-3-fluoro trifluoromethoxy benzene, 3-fluoro-4-bromobenzenetrifluoromethoxy, 4-bromo-3-fluoro-trifluoromethoxy benzene, benzene, 1-bromo-2-fluoro-4-trifluoromethoxy, pubchem1075, acmc-1c50f, ksc498c3b, 4-bromo-3-fluorotrifluoromethoxy-benzene, 2-fluoro-4-trifluoromethoxy bromobenzene
1-bromo-2-fluoro-4-(trifluoromethoxy)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents