Please login to your online account to display your discounted pricing

1-Bromo-2-fluoro-4-iodobenzene, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-fluoro-4-iodobenzene
CAS 136434-77-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H3BrFI
Linear Formula BrC6H3(F)I
Formula Weight 300.89
MDL Number MFCD00042577
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC295430050

  SDS
thermo fisher scientific
AC295430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Crystalline Low Melting Solid
Formula Weight 300.89
Quantity 5g
Color Pale brown
Melting Point 36° to 41°C
CAS Min % 98.0
CAS Max % 100.0
Linear Formula BrC6H3(F)I
Chemical Name or Material 1-Bromo-2-fluoro-4-iodobenzene
Assay Percent Range 98+%
Molecular Formula C6H3BrFI
MDL Number MFCD00042577
CAS 136434-77-0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic