Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-(difluoromethoxy)benzene, 97+%

$118.58 - $423.41

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-(difluoromethoxy)benzene
CAS 175278-33-8
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD00221468
Synonym 2-(Difluoromethoxy)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2317404

  SDS
alfa aesar™
B2317404
2g Each for $118.58

AAB2317409

  SDS
alfa aesar™
B2317409
10g Each for $423.41
Description & Specifications

Specifications

Formula Weight 223.02
Health Hazard 1 Warning
Molecular Formula C7H5BrF2O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 175278-33-8
Chemical Name or Material 1-Bromo-2-(difluoromethoxy)benzene
Boiling Point 85° to 87°C (15mmHg)
Synonym 2-(Difluoromethoxy)bromobenzene
Assay ≥97%
Quantity 2g
MDL Number MFCD00221468
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.