missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-cyclohexylbenzene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1805906

 View more versions of this product

Catalog No. AAL1805906



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-2-cyclohexylbenzene
1.295
C12H15Br
MFCD02093987
2365764
BXMDAGGQRIDSPA-UHFFFAOYSA-N
1-bromo-2-cyclohexylbenzene
10944502
97%
59734-92-8
79°C to 83°C (0.01 mmHg)
1.5605
5 g
1-bromo-2-cyclohexyl-benzene, benzene, 1-bromo-2-cyclohexyl, acmc-20aoxa, l-bromo-2-cyclohexylbenzene, 2-bromo-1-cyclohexylbenzene
BrC1=CC=CC=C1C1CCCCC1
239.16
239.16

Chemical Identifiers

59734-92-8
239.16
BXMDAGGQRIDSPA-UHFFFAOYSA-N
10944502
BrC1=CC=CC=C1C1CCCCC1
C12H15Br
MFCD02093987
1-bromo-2-cyclohexyl-benzene, benzene, 1-bromo-2-cyclohexyl, acmc-20aoxa, l-bromo-2-cyclohexylbenzene, 2-bromo-1-cyclohexylbenzene
1-bromo-2-cyclohexylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only