Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-chlorotetrafluoroethane, 98%

$350.96 - $350.96

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-chlorotetrafluoroethane
CAS 354-53-0
Molecular Formula C2BrClF4
Linear Formula ClCF2CF2Br
Formula Weight 215.37
MDL Number MFCD00042179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2042709

 
alfa aesar™
B2042709
10g Each for $350.96
Description & Specifications

Specifications

Formula Weight 215.37
Chemical Name or Material 1-Bromo-2-chlorotetrafluoroethane
Assay 98%
Molecular Formula C2BrClF4
Linear Formula ClCF2CF2Br
MDL Number MFCD00042179
CAS 354-53-0
Melting Point −123°C
Boiling Point 25°C
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C