missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-chloroethane, 98%

Manufacturer:  Alfa Aesar™ A1234922*

 View more versions of this product

Catalog No. AAA1234922



Description

Description

Applications
1-Bromo-2-chloroethane was used in regio- and diastereoselective synthesis of 2-alkylidenetetrahydrofurans. 1-Bromo-2-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigant.

Solubility
Soluble in water ( 7 g/L (20°C). Soluble in ethanol, ethyl ether, chloroform.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

1-Bromo-2-chloroethane
106°C to 107°C
100g
C2H4BrCl
MFCD00000962
605265
IBYHHJPAARCAIE-UHFFFAOYSA-N
1-bromo-2-chloroethane
7849
143.42
Sweet
-18°C
107-04-0
ClCH2CH2Br
UN2810
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
C(CBr)Cl
143.408
CHEBI:19032
98%

Chemical Identifiers

107-04-0
143.408
IBYHHJPAARCAIE-UHFFFAOYSA-N
7849
1-bromo-2-chloroethane
C2H4BrCl
MFCD00000962
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
CHEBI:19032
C(CBr)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H350-H312-H332-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : KH6500000

TSCA : TSCA

SDS Documents