missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-chlorobenzene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1178218

 View more versions of this product

Catalog No. AAA1178218



Description

Description

1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-chlorobenzene finds application in Gas Chromatography and Liquid Chromatography analysis. It is also used as an intermediate in pharmaceutical and synthetic chemistry. Further, it acts as a reagent in the preparation of arylpiperidines and aryltetrahydropyridines as 5-HT2C agonists.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

1-Bromo-2-chlorobenzene
694-80-4
203°C to 205°C
1.5815
MFCD00000532
2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene
QBELEDRHMPMKHP-UHFFFAOYSA-N
1-bromo-2-chlorobenzene
12754
≥98%
-13°C
1.649
50 g
C6H4BrCl
1855303
Immiscible with water.
C1=CC=C(C(=C1)Cl)Br
191.452
191.46

Chemical Identifiers

694-80-4
191.452
QBELEDRHMPMKHP-UHFFFAOYSA-N
12754
C1=CC=C(C(=C1)Cl)Br
C6H4BrCl
MFCD00000532
2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene
1-bromo-2-chlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H332-H315-H319-H335

EINECSNumber : 211-775-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only