Please login to your online account to display your discounted pricing

1-Bromo-2-butyne 98%, ACROS Organics™


Quantity 25g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC387020250

 View more versions of this product

Catalog No. AC387020250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear pale yellow-greenish
Physical Form Liquid
Assay ≥97.5%
Formula Weight 132.99
Quantity 25g
Synonym 2-Butynyl Bromide
Packaging Glass Bottle
Density 1.4730g/mL
Boiling Point 40.0° to 41.0°C
Flash Point 36°C
UN Number 1993
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C4H5Br
Linear Formula CH3CCCH2Br
MDL Number MFCD00190233
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Refractive Index 1.5070 to 1.5090 (20°C, 589nm)
CAS 3355-28-0
Infrared Spectrum Authentic
Chemical Name or Material 1-Bromo-2-butyne
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Bromo-2-butyne
CAS 3355-28-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H5Br
Linear Formula CH3CCCH2Br
Formula Weight 132.99
MDL Number MFCD00190233
Synonym 2-Butynyl Bromide
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents