Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2-butanone, tech. 85%, stab. with calcium carbonate

Chemical Identifiers

Chemical Name or Material 1-Bromo-2-butanone
CAS 816-40-0
Name Note stabilized with calcium carbonate
Purity Grade Technical Grade
Molecular Formula C4H7BrO
Formula Weight 151.01g/mol
MDL Number MFCD00000207
UN Number UN2927
Beilstein 741894
Synonym Bromomethyl ethyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1208103

 
alfa aesar™
L12081-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1208106

 
alfa aesar™
L12081-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Boiling Point 48° to 49°C (10mmHg)
Quantity 1g
Flash Point 68°C (154°F)
Odor Pungent
Chemical Name or Material 1-Bromo-2-butanone
Name Note stabilized with calcium carbonate
CAS 816-40-0
Purity Grade Technical Grade
Assay 85%
Molecular Formula C4H7BrO
Formula Weight 151.01g/mol
MDL Number MFCD00000207
UN Number UN2927
Beilstein 741894
Synonym Bromomethyl ethyl ketone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
RTECS Number EL7000000
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.