Please login to your online account to display your discounted pricing

1-Bromo-2,6-difluorobenzene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-2, 6-difluorobenzene
CAS 64248-56-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3BrF2
Linear Formula F2C6H3Br
Formula Weight 192.99
MDL Number MFCD00009894
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC297390010

  SDS
thermo fisher scientific
AC297390010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 192.99
Physical Form Liquid
Quantity 1g
Color Clear yellow
Density 1.7100g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula F2C6H3Br
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
CAS 64248-56-2
Chemical Name or Material 1-Bromo-2, 6-difluorobenzene
Molecular Formula C6H3BrF2
Assay Percent Range 98%
MDL Number MFCD00009894
CAS Min % 97.5
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.