missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,6-dichlorobenzene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 171040250

 View more versions of this product

Catalog No. AC171040250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
225.9
White to yellow
62° to 66°C
Glass bottle
C6H3BrCl2
19393-92-1
1-Bromo-2, 6-dichlorobenzene
99%
Crystalline Powder and/or Chunks
25g
05, 210
100.0
Authentic
MFCD00000574
BrC6H3Cl2
98.5
99%

Chemical Identifiers

19393-92-1
100.0
C6H3BrCl2
225.9
1-Bromo-2, 6-dichlorobenzene, 99%
98.5
99%
BrC6H3Cl2
MFCD00000574
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-018-6

TSCA : TSCA