missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,6-dichlorobenzene, 99%, ACROS Organics™

Manufacturer:  Acros Organics 171040050

 View more versions of this product

Catalog No. AC171040050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Bromo-2, 6-dichlorobenzene
Glass bottle
White to Yellow
5g
19393-92-1
100.0
C6H3BrCl2
MFCD00000574
1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
C1=CC(=C(C(=C1)Cl)Br)Cl
225.894
225.9
99%
Authentic
242.0°C (765.0 mmHg)
64.0°C to 67.0°C
99%
98.5
99%
BrC6H3Cl2
05, 210
UWOIDOQAQPUVOH-UHFFFAOYSA-N
2-bromo-1,3-dichlorobenzene
29568
Crystalline Powder and/or Chunks

Chemical Identifiers

19393-92-1
225.894
UWOIDOQAQPUVOH-UHFFFAOYSA-N
29568
C1=CC(=C(C(=C1)Cl)Br)Cl
C6H3BrCl2
MFCD00000574
1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
2-bromo-1,3-dichlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-018-6

TSCA : TSCA

SDS Documents