Please login to your online account to display your discounted pricing

1-Bromo-2,6-dichlorobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-2, 6-dichlorobenzene
Name Note 99%
CAS 19393-92-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3BrCl2
Linear Formula BrC6H3Cl2
Formula Weight 225.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171040050

  SDS
thermo fisher scientific
171040050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171040250

  SDS
thermo fisher scientific
171040250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder and/or Chunks
Formula Weight 225.9
Quantity 5g
Color White to yellow
EINECS Number 243-018-6
TSCA TSCA
Melting Point 62° to 66°C
Beilstein 05, 210
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Bromo-2, 6-dichlorobenzene
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H3BrCl2
Linear Formula BrC6H3Cl2
MDL Number MFCD00000574
CAS 19393-92-1
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.