Please login to your online account to display your discounted pricing

1-Bromo-2,5-difluorobenzene, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  249370010

 View more versions of this product

Catalog No. AC249370010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 192.99
Physical Form Liquid
Quantity 1g
Color Clear colorless
EINECS Number 206-920-0
TSCA TSCA
CAS Max % 100.0
Refractive Index 1.5075 to 1.5095 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 98%
Chemical Name or Material 1-Bromo-2, 5-difluorobenzene
CAS 399-94-0
MDL Number MFCD00000345
CAS Min % 97.5
Molecular Formula C6H3BrF2
Linear Formula FC6H3BrF
Assay Percent Range 98%
Packaging Glass bottle
Synonym 2-Bromo-1, 4-difluorobenzene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Bromo-2, 5-difluorobenzene
Name Note 98%
CAS 399-94-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3BrF2
Linear Formula FC6H3BrF
Formula Weight 192.99
MDL Number MFCD00000345
Synonym 2-Bromo-1, 4-difluorobenzene
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents