Please login to your online account to display your discounted pricing

1-Bromo-2,5-difluorobenzene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-2, 5-difluorobenzene
Name Note 98%
CAS 399-94-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3BrF2
Linear Formula FC6H3BrF
Formula Weight 192.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC249370010

  SDS
thermo fisher scientific
249370010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC249370050

  SDS
thermo fisher scientific
249370050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 192.99
Quantity 1g
Color Clear colorless
EINECS Number 206-920-0
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 1-Bromo-2, 5-difluorobenzene
Refractive Index 1.5075 to 1.5095 (20°C, 589nm)
Name Note 98%
MDL Number MFCD00000345
CAS 399-94-0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H3BrF2
Linear Formula FC6H3BrF
Synonym 2-Bromo-1, 4-difluorobenzene
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.