Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,5-difluoro-4-nitrobenzene, 97%

$107.30 - $387.89

Chemical Identifiers

Chemical Name or Material 1-Bromo-2,5-difluoro-4-nitrobenzene
CAS 167415-27-2
Molecular Formula C6H2BrF2NO2
Formula Weight 237.99
MDL Number MFCD00042184
Beilstein 7411347
Synonym 4-Bromo-2,5-difluoronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1867003

  SDS
alfa aesar™
A1867003
1g Each for $107.30

AAA1867006

  SDS
alfa aesar™
A1867006
5g Each for $387.89
Description & Specifications

Specifications

Formula Weight 237.99
Chemical Name or Material 1-Bromo-2,5-difluoro-4-nitrobenzene
Assay 97%
Molecular Formula C6H2BrF2NO2
MDL Number MFCD00042184
CAS 167415-27-2
Melting Point 55° to 57°C
Beilstein 7411347
Synonym 4-Bromo-2,5-difluoronitrobenzene
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.