Please login to your online account to display your discounted pricing

1-Bromo-2,5-dichlorobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Bromo-2, 5-dichlorobenzene
Name Note 99%
CAS 1435-50-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3BrCl2
Formula Weight 225.9
MDL Number MFCD00018505
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC290720050

  SDS
thermo fisher scientific
290720050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Chunks
Assay ≥98.5%
Formula Weight 225.9
Quantity 5g
Color Pale yellow to beige
EINECS Number 215-859-9
TSCA TSCA
Melting Point 31° to 35°C
CAS Max % 100.0
Chemical Name or Material 1-Bromo-2, 5-dichlorobenzene
Assay Percent Range 99%
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Molecular Formula C6H3BrCl2
MDL Number MFCD00018505
CAS 1435-50-3
Synonym 2-Bromo-1, 4-dichlorobenzene
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.