Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,5-bis(trifluoromethyl)benzene, 97%

$46.09 - $158.22

Chemical Identifiers

Chemical Name or Material 1-Bromo-2,5-bis(trifluoromethyl)benzene
CAS 7617-93-8
Molecular Formula C8H3BrF6
Formula Weight 293.01
MDL Number MFCD03094336
Synonym 2,5-Bis(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2549103

  SDS
alfa aesar™
B2549103
1g Each for $46.09

AAB2549106

  SDS
alfa aesar™
B2549106
5g Each for $158.22
Description & Specifications

Specifications

Formula Weight 293.01
Chemical Name or Material 1-Bromo-2,5-bis(trifluoromethyl)benzene
Assay 97%
Molecular Formula C8H3BrF6
MDL Number MFCD03094336
CAS 7617-93-8
Synonym 2,5-Bis(trifluoromethyl)bromobenzene
Boiling Point 146° to 147°C
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.