missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,4-dimethyl-5-nitrobenzene, 95%

$44.86 - $661.57

Chemical Identifiers

CAS 69383-59-1
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00075824
InChI Key HYYJANCKMGXHLQ-UHFFFAOYSA-N
Synonym 5-bromo-2,4-dimethylnitrobenzene, 4-bromo-6-nitro-m-xylene, benzene, 1-bromo-2,4-dimethyl-5-nitro, 1-bromo-2,4-dimethyl-5-nitro-benzene, 2-nitro-4-bromo-m-xylene, 2,4-dimethyl-5-nitro-1-bromobenzene, pubchem19926, benzene,1-bromo-2,4-dimethyl-5-nitro
PubChem CID 12474326
IUPAC Name 1-bromo-2,4-dimethyl-5-nitrobenzene
SMILES CC1=CC(=C(C=C1[N+](=O)[O-])Br)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6620803 View Documents Alfa Aesar™
H6620803
1g Each for $44.86
 
AAH6620806 View Documents Alfa Aesar™
H6620806
5g Each for $166.55
 
AAH6620814 View Documents Alfa Aesar™
H6620814
25g Each for $661.57
 
Description

Description

Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

69383-59-1
230.061
HYYJANCKMGXHLQ-UHFFFAOYSA-N
12474326
CC1=CC(=C(C=C1[N+](=O)[O-])Br)C
C8H8BrNO2
MFCD00075824
5-bromo-2,4-dimethylnitrobenzene, 4-bromo-6-nitro-m-xylene, benzene, 1-bromo-2,4-dimethyl-5-nitro, 1-bromo-2,4-dimethyl-5-nitro-benzene, 2-nitro-4-bromo-m-xylene, 2,4-dimethyl-5-nitro-1-bromobenzene, pubchem19926, benzene,1-bromo-2,4-dimethyl-5-nitro
1-bromo-2,4-dimethyl-5-nitrobenzene

Specifications

Yellow
69383-59-1
MFCD00075824
1948547
HYYJANCKMGXHLQ-UHFFFAOYSA-N
1-bromo-2,4-dimethyl-5-nitrobenzene
12474326
Solid
1-Bromo-2,4-dimethyl-5-nitrobenzene
1g
C8H8BrNO2
UN2811
5-bromo-2,4-dimethylnitrobenzene, 4-bromo-6-nitro-m-xylene, benzene, 1-bromo-2,4-dimethyl-5-nitro, 1-bromo-2,4-dimethyl-5-nitro-benzene, 2-nitro-4-bromo-m-xylene, 2,4-dimethyl-5-nitro-1-bromobenzene, pubchem19926, benzene,1-bromo-2,4-dimethyl-5-nitro
CC1=CC(=C(C=C1[N+](=O)[O-])Br)C
230.061
230.06
95%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C