missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2,4-dimethoxybenzene, 98%, ACROS Organics™

$142.27 - $142.27

Chemical Identifiers

CAS 17715-69-4
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD00009844
InChI Key NIUZVSQOXJIHBL-UHFFFAOYSA-N
Synonym 1,3-dimethoxy-4-bromobenzene, 1-brom-2,4-dimethoxybenzol, 2,4-dimethoxybromobenzene, 4-bromo-1,3-dimethoxybenzene, 4-bromo-3-methoxyanisole, 4-bromoresocinol dimethyl ether, 4-bromoresorcinol dimethyl ether, acmc-1c5dw, benzene, 1-bromo-2,4-dimethoxy, pubchem3036
PubChem CID 87266
IUPAC Name 1-bromo-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)Br)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC134170050
SDS View Documents
Thermo Fisher Scientific
134170050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC134170250
SDS View Documents
Thermo Fisher Scientific
134170250
25mL Glass bottle
Each for $142.27
 
Description

Description

Specifications

Chemical Identifiers

17715-69-4
217.06
NIUZVSQOXJIHBL-UHFFFAOYSA-N
87266
COC1=CC(=C(C=C1)Br)OC
C8H9BrO2
MFCD00009844
1,3-dimethoxy-4-bromobenzene, 1-brom-2,4-dimethoxybenzol, 2,4-dimethoxybromobenzene, 4-bromo-1,3-dimethoxybenzene, 4-bromo-3-methoxyanisole, 4-bromoresocinol dimethyl ether, 4-bromoresorcinol dimethyl ether, acmc-1c5dw, benzene, 1-bromo-2,4-dimethoxy, pubchem3036
1-bromo-2,4-dimethoxybenzene

Specifications

1.5070g/mL
Authentic
1.5718 to 1.5738
153°C to 155°C (18.0mmHg)
25°C to 26°C
17715-69-4
100.0
C8H9BrO2
MFCD00009844
1,3-dimethoxy-4-bromobenzene, 1-brom-2,4-dimethoxybenzol, 2,4-dimethoxybromobenzene, 4-bromo-1,3-dimethoxybenzene, 4-bromo-3-methoxyanisole, 4-bromoresocinol dimethyl ether, 4-bromoresorcinol dimethyl ether, acmc-1c5dw, benzene, 1-bromo-2,4-dimethoxy, pubchem3036
NIUZVSQOXJIHBL-UHFFFAOYSA-N
1-bromo-2,4-dimethoxybenzene
87266
Liquid
1-Bromo-2, 4-dimethoxybenzene, 98%
109°C
Glass bottle
1.507
Yellow
5mL
97.5
97.5% min. (GC)
CH3OC6H3(OCH3)Br
06, II, 819
Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
COC1=CC(=C(C=C1)Br)OC
217.06
217.06
98%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-717-0

TSCA : TSCA

SDS