Please login to your online account to display your discounted pricing

1-Bromo-2,4-dimethoxybenzene 98%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  134170250

 View more versions of this product

Catalog No. AC134170250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 217.06
Physical Form Liquid
Assay ≥97.5%
Quantity 25mL
Color Clear slightly yellow
Beilstein 06, II, 819
TSCA TSCA
EINECS Number 241-717-0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5718 to 1.5738 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Bromo-2, 4-dimethoxybenzene
Linear Formula CH3OC6H3(OCH3)Br
MDL Number MFCD00009844
CAS Min % 97.5
Molecular Formula C8H9BrO2
CAS 17715-69-4
Name Note 98%
Assay Percent Range 98%
Infrared Spectrum Authentic
Synonym 4-Bromoresorcinol dimethylether

Specifications

Chemical Name or Material 1-Bromo-2, 4-dimethoxybenzene
Name Note 98%
CAS 17715-69-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9BrO2
Linear Formula CH3OC6H3(OCH3)Br
Formula Weight 217.06
MDL Number MFCD00009844
Synonym 4-Bromoresorcinol dimethylether
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents