Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Bromo-2,4-bis(trifluoromethyl)benzene, 97%

$50.18 - $173.28

Chemical Identifiers

Chemical Name or Material 1-Bromo-2,4-bis(trifluoromethyl)benzene
CAS 327-75-3
Molecular Formula C8H3BrF6
Formula Weight 293.01
MDL Number MFCD00074904
Beilstein 6208648
Synonym 2,4-Bis(trifluoromethyl)bromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2463003

  SDS
alfa aesar™
B2463003
1g Each for $50.18

AAB2463006

  SDS
alfa aesar™
B2463006
5g Each for $173.28
Description & Specifications

Specifications

Formula Weight 293.01
Chemical Name or Material 1-Bromo-2,4-bis(trifluoromethyl)benzene
Quantity 1g
Health Hazard 1 Warning
Synonym 2,4-Bis(trifluoromethyl)bromobenzene
Assay 97%
CAS 327-75-3
Boiling Point 163°C
Beilstein 6208648
Molecular Formula C8H3BrF6
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00074904
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.